EMAIL THIS PAGE TO A FRIEND
Nature communications
A mast cell-ILC2-Th9 pathway promotes lung inflammation in cystic fibrosis.
PMID 28090087
Abstract
T helper 9 (Th9) cells contribute to lung inflammation and allergy as sources of interleukin-9 (IL-9). However, the mechanisms by which IL-9/Th9 mediate immunopathology in the lung are unknown. Here we report an IL-9-driven positive feedback loop that reinforces allergic inflammation. We show that IL-9 increases IL-2 production by mast cells, which leads to expansion of CD25