EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Synthesis, pH dependent photometric and electrochemical investigation, redox mechanism and biological applications of novel Schiff base and its metallic derivatives.
PMID 28092826
Abstract
A novel Schiff base, 1-((2, 4-dimethylphenylimino)methyl)naphthalen-2-ol abbreviated as (HL) and its four metallic complexes were synthesized and confirmed by
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H45353
2-Hydroxy-1-naphthaldehyde, technical grade
C11H8O2
205494
Dibutyltin dichloride, 96%
C8H18Cl2Sn