EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Conjugated polymer with carboxylate groups-Hg
PMID 28092827
Abstract
In this work, a turn on fluorescent sensor, based on Hg
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
632090
1,4-Diethynylbenzene, 96%
C10H6
216666
Tetrakis(triphenylphosphine)palladium(0), 99%
C72H60P4Pd