EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
LC-MS/MS determination of d-mannose in human serum as a potential cancer biomarker.
PMID 28092855
Abstract
Several metabolites in human serum have been identified as potential cancer biomarkers for early detection. This study focuses on the LC-MS/MS method development and validation of d-mannose in human serum. Surrogate blank serum, coupled with stable isotope d-mannose-
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
63579
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
C6H12O6