EMAIL THIS PAGE TO A FRIEND
Analytica chimica acta
Hairpin stabilized fluorescent silver nanoclusters for quantitative detection of NAD
PMID 28093125
Abstract
A set of 90 mer long ssDNA candidates, with different degrees of cytosine (C-levels) (% and clusters) was analyzed for their function as suitable Ag-nanocluster (AgNC) nucleation scaffolds. The sequence (P4) with highest C-level (42.2%) emerged as the only candidate supporting the nucleation process as evident from its intense fluorescence peak at λ