EMAIL THIS PAGE TO A FRIEND
Physical chemistry chemical physics : PCCP
Effect of electron spectral diffusion on static dynamic nuclear polarization at 7 Tesla.
PMID 28094364
Abstract
Here, we present an integrated experimental and theoretical study of
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
163945
4-Amino-TEMPO, free radical, 97%
C9H19N2O