EMAIL THIS PAGE TO A FRIEND
Nano letters
Spatially Resolved Imaging on Photocarrier Generations and Band Alignments at Perovskite/PbI
PMID 28094957
Abstract
The presence of the PbI
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
284513
Chlorobenzene, anhydrous, 99.8%
C6H5Cl