EMAIL THIS PAGE TO A FRIEND
Cell & bioscience
A fission yeast cell-based system for multidrug resistant HIV-1 proteases.
PMID 28096973
Abstract
HIV-1 protease (PR) is an essential enzyme for viral production. Thus, PR inhibitors (PIs) are the most effective class of anti-HIV drugs. However, the main challenge to the successful use of PI drugs in patient treatment is the emergence of multidrug resistant PRs ( Three The three clinically isolated
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
69697
1-Methylpyridinium iodide, analytical standard
C6H8IN