EMAIL THIS PAGE TO A FRIEND
Journal of fluorescence
Synthesis, Spectral Characterization, DNA/ Protein Binding, DNA Cleavage, Cytotoxicity, Antioxidative and Molecular Docking Studies of Cu(II)Complexes Containing Schiff Base-bpy/Phen Ligands.
PMID 28097460
Abstract
Ternary Cu(II) complexes [Cu(II)(L)(bpy)Cl] 1, [Cu(II)(L)(Phen)Cl] 2 [L = 2,3-dimethyl-1-phenyl-4(2 hydroxy-5-methyl benzylideneamino)-pyrazol-5-one, bpy = 2,2
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
454664
2-Hydroxy-5-methylbenzaldehyde, 98%
C8H8O2