EMAIL THIS PAGE TO A FRIEND
Endocrine
Selenium rescues orbital fibroblasts from cell death induced by hydrogen peroxide: another molecular basis for the effects of selenium in graves' orbitopathy.
PMID 28097622