EMAIL THIS PAGE TO A FRIEND
Nature communications
Carbon translocation from a plant to an insect-pathogenic endophytic fungus.
PMID 28098142
Abstract
Metarhizium robertsii is a common soil fungus that occupies a specialized ecological niche as an endophyte and an insect pathogen. Previously, we showed that the endophytic capability and insect pathogenicity of Metarhizium are coupled to provide an active method of insect-derived nitrogen transfer to a host plant via fungal mycelia. We speculated that in exchange for this insect-derived nitrogen, the plant would provide photosynthate to the fungus. By using