EMAIL THIS PAGE TO A FRIEND
Nature communications
Expression of the novel maternal centrosome assembly factor Wdr8 is required for vertebrate embryonic mitoses.
PMID 28098238
Abstract
The assembly of the first centrosome occurs upon fertilisation when male centrioles recruit pericentriolar material (PCM) from the egg cytoplasm. The mechanisms underlying the proper assembly of centrosomes during early embryogenesis remain obscure. We identify Wdr8 as a novel maternally essential protein that is required for centrosome assembly during embryonic mitoses of medaka (Oryzias latipes). By CRISPR-Cas9-mediated knockout, maternal/zygotic Wdr8-null (m/zWdr8
Related Materials