EMAIL THIS PAGE TO A FRIEND
Expert opinion on pharmacotherapy
New and developing non-adrenoreceptor small molecule drugs for the treatment of asthma.
PMID 28099820
Abstract
Inhaled corticosteroids (ICS) alone or in combination with an inhaled long-acting beta
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML2008
Tiotropium bromide monohydrate, ≥98% (HPLC)
C19H22BrNO4S2 · H2O