EMAIL THIS PAGE TO A FRIEND
PMID 28100772
Abstract
Sphingolipids are a diverse class of essential cellular lipids that function as structural membrane components and as signaling molecules. Cells acquire sphingolipids by both
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
12094
Docosanoic acid tryptamide, analytical standard
C32H54N2O