EMAIL THIS PAGE TO A FRIEND
International journal of oncology
The protein arginine methyltransferase 5 promotes malignant phenotype of hepatocellular carcinoma cells and is associated with adverse patient outcomes after curative hepatectomy.
PMID 28101581
Abstract
The prognosis of advanced hepatocellular carcinoma (HCC) is dismal. Novel molecular targets for diagnosis and therapy is urgently required. This study evaluated expression and functions of the protein arginine methyltransferase 5 (PRMT5) in HCC. Using HCC cell lines, the expression levels of PRMT5 mRNA were determined using the quantitative real-time reverse-transcription polymerase chain reaction, and the effect of a small interfering PRMT5-siRNA on cell phenotype was evaluated. Further, PRMT5 expression was determined in 144 pairs of resected liver tissues to evaluate its clinical significance. Regardless of their differentiated phenotypes, nine HCC cell lines expressed different levels of PRMT5 mRNA. Inhibition of PRMT5 expression significantly decreased the proliferation, invasion, and migration of HCC cell lines. Although the level of PRMT5 mRNA was not influenced by patient's background liver status, it was significantly higher in HCC tissues than in the corresponding noncancerous tissues. High levels of PRMT5 mRNA in HCC tissues were significantly associated with advanced disease stage and adverse prognosis. In conclusion, our results indicate that PRMT5 may act as a putative oncogene in HCC and that the levels of PRMT5 mRNA represent a promising prognostic marker and a potential target of molecular therapy for HCC.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU065921 MISSION® esiRNA, esiRNA human PRMT5 (esiRNA1)