EMAIL THIS PAGE TO A FRIEND
Journal of proteome research
Deep Coverage of Global Protein Expression and Phosphorylation in Breast Tumor Cell Lines Using TMT 10-plex Isobaric Labeling.
PMID 28102081
Abstract
Labeling peptides with isobaric tags is a popular strategy in quantitative bottom-up proteomics. In this study, we labeled six breast tumor cell lysates (1.34 mg proteins per channel) using 10-plex tandem mass tag reagents and analyzed the samples on a Q Exactive HF Quadrupole-Orbitrap mass spectrometer. We identified a total of 8,706 proteins and 28,186 phosphopeptides, including 7,394 proteins and 23,739 phosphosites common to all channels. The majority of technical replicates correlated with a R