EMAIL THIS PAGE TO A FRIEND
Neuron
Synaptic Plasticity onto Dopamine Neurons Shapes Fear Learning.
PMID 28103482
Abstract
Fear learning is a fundamental behavioral process that requires dopamine (DA) release. Experience-dependent synaptic plasticity occurs on DA neurons while an organism is engaged in aversive experiences. However, whether synaptic plasticity onto DA neurons is causally involved in aversion learning is unknown. Here, we show that a stress priming procedure enhances fear learning by engaging VTA synaptic plasticity. Moreover, we took advantage of the ability of the ATPase Thorase to regulate the internalization of AMPA receptors (AMPARs) in order to selectively manipulate glutamatergic synaptic plasticity on DA neurons. Genetic ablation of Thorase in DAT
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D0540
DNQX, ≥98% (TLC)
C8H4N4O6