EMAIL THIS PAGE TO A FRIEND
Colloids and surfaces. B, Biointerfaces
Topical vaccination with super-stable ready to use nanovesicles.
PMID 28103528
Abstract
Ultradeformable archaeosomes (UDA) are nanovesicles made of total polar archaeolipids (TPA) from the archaea Halorubrum tebenquichense, soybean phosphatidylcholine and sodium cholate (3:3:1w/w). Fresh dispersions of UDA including different type of antigens are acknowledged as efficient topical vaccination agents. UDA dispersions however, if manufactured for pharmaceutical use, have to maintain colloidal stability upon liposomicidal processes such as sterilization and lyophilization (
Related Materials