EMAIL THIS PAGE TO A FRIEND
Experimental and therapeutic medicine
Oxidized low-density lipoprotein decreases VEGFR2 expression in HUVECs and impairs angiogenesis.
PMID 28105106
Abstract
Atherosclerosis (AS), which is triggered by endothelial cell injury, evolves into a chronic inflammatory disease. Oxidized low-density lipoprotein (ox-LDL) is an important risk factor for the development of atherosclerosis; ox-LDL induces atherosclerotic plaque formation via scavenging receptors. The present study used ox-LDL-treated human umbilical vein endothelial cells (HUVECs) to investigate the effect of ox-LDL on angiogenesis. ox-LDL decreased HUVEC proliferation by MTT, induced apoptosis by Annexin V-fluorescein isothiocyanate (FITC) staining and markedly suppressed HUVEC tube formation by the Matrigel assay in a dose-dependent manner. Angiogenesis has been correlated with monocyte invasion, plaque instability and atherosclerotic lesion formation. In addition, ox-LDL induced the overproduction of reactive oxygen species using DCFH-DA staining and increased caspase-3 activity. Vascular endothelial growth factor receptor 2 (VEGFR2) were detected by quantitative polymerase chain reaction and western blot analysis and has previously been observed to have a key role in angiogenesis. Furthermore, the present study demonstrated that the abundance of VEGFR2 was decreased in ox-LDL-treated HUVECs. These results suggested that ox-LDL impairs angiogenesis via VEGFR2 degradation, thus suggesting that VEGFR2 may be involved in adaptation to oxidative stress and AS.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2108266
Anti-GAPDH antibody produced in rabbit, affinity isolated antibody