Journal of cardiovascular translational research
Phenotype-Specific Association of Single-Nucleotide Polymorphisms with Heart Failure and Preserved Ejection Fraction: a Genome-Wide Association Analysis of the Cardiovascular Health Study.
PMID 28105587
Little is known about genetics of heart failure with preserved ejection fraction (HFpEF) in part because of the many comorbidities in this population. To identify single-nucleotide polymorphisms (SNPs) associated with HFpEF, we analyzed phenotypic and genotypic data from the Cardiovascular Health Study, which profiled patients using a 50,000 SNP array. Results were explored using novel SNP- and gene-centric tools. We performed analyses to determine whether some SNPs were relevant only in certain phenotypes. Among 3804 patients, 7 clinical factors and 9 SNPs were significantly associated with HFpEF; the most notable of which was rs6996224, a SNP associated with transforming growth factor-beta receptor 3. Most SNPs were associated with HFpEF only in the absence of a clinical predictor. Significant SNPs represented genes involved in myocyte proliferation, transforming growth factor-beta/erbB signaling, and extracellular matrix formation. These findings suggest that genetic factors may be more important in some phenotypes than others.