PloS one
Studies on New Activities of Enantiomers of 2-(2-Hydroxypropanamido) Benzoic Acid: Antiplatelet Aggregation and Antithrombosis.
PMID 28107496
Abstract
R-/S-2-(2-Hydroxypropanamido) benzoic acid (R-/S-HPABA), a marine-derived anti-inflammatory drug, however, the antiplatelet and antithrombotic effects have not been investigated. In this paper, the in vitro antiplatelet activities and in vivo antithrombotic effects of R-/S-HPABA were investigated, for the first time. The effects of R-/S-HPABA on platelet aggregation induced by adenosine diphosphate (ADP), collagen (COLL) and arachidonic acid (AA) were evaluated. In addition, the in vivo bleeding time, clotting time, collagen-epinephrine induced pulmonary thrombosis and common carotid artery thrombosis were also investigated in rats. R-/S-HPABA significantly inhibited ADP, COLL and AA induced platelet aggregation in rabbit platelet rich plasma in vitro compared with control group, to a degree similar to that of aspirin. Besides, R-/S-HPABA prolonged bleeding time and clotting time as well as increased the recovery rate obviously in pulmonary thrombosis. Moreover, the level of thromboxane B2 (TXB2) was decreased while the production of 6-keto-prostaglandin F1α (6-keto-PGF1α) was increased markedly by R-/S-HPABA. Furthermore, R-/S-HPABA reduced carotid artery thrombosis weight. These results illustrated that R-/S-HPABA could be a potent antiplatelet aggregation and antithrombotic agent.