EMAIL THIS PAGE TO A FRIEND
ACS nano
Gold Nanoshell-Mediated Remote Myotube Activation.
PMID 28107625
Abstract
Mild heat stimulation of muscle cells within the physiological range represents an intriguing approach for the modulation of their functions. In this work, photothermal conversion was exploited to remotely stimulate striated muscle cells by using gold nanoshells (NSs) in combination with near-infrared (NIR) radiation. Temperature increments of approximately 5 °C were recorded by using an intracellular fluorescent molecular thermometer and were demonstrated to efficiently induce myotube contraction. The mechanism at the base of this phenomenon was thoroughly investigated and was observed to be a Ca
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
793728
1,3,5,7-Tetramethyl-8-phenyl-4,4-difluoroboradiazaindacene, 97%
C19H19BF2N2