EMAIL THIS PAGE TO A FRIEND
International journal for parasitology. Drugs and drug resistance
Effect of clinically approved HDAC inhibitors on Plasmodium, Leishmania and Schistosoma parasite growth.
PMID 28107750
Abstract
Malaria, schistosomiasis and leishmaniases are among the most prevalent tropical parasitic diseases and each requires new innovative treatments. Targeting essential parasite pathways, such as those that regulate gene expression and cell cycle progression, is a key strategy for discovering new drug leads. In this study, four clinically approved anti-cancer drugs (Vorinostat, Belinostat, Panobinostat and Romidepsin) that target histone/lysine deacetylase enzymes were examined for in vitro activity against Plasmodium knowlesi, Schistosoma mansoni, Leishmania amazonensis and L. donovani parasites and two for in vivo activity in a mouse malaria model. All four compounds were potent inhibitors of P. knowlesi malaria parasites (IC
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML1175
Romidepsin, ≥98% (HPLC)
C24H36N4O6S2