EMAIL THIS PAGE TO A FRIEND
Talanta
Novel potentiometric sensors for the determination of the dinotefuran insecticide residue levels in cucumber and soil samples.
PMID 28107966
Abstract
Five new potentiometric membrane sensors for the determination of the dinotefuran levels in cucumber and soil samples have been developed. Four of these sensors were based on a newly designed molecularly imprinted polymer (MIP) material consisting of acrylamide or methacrylic acid as a functional monomer in a plasticized PVC (polyvinyl chloride) membrane before and after elution of the template. A fifth sensor, a carboxylated PVC-based sensor plasticized with dioctyl phthalate, was also prepared and tested. Sensor 1 (acrylamide washed) and sensor 3 (methacrylic acid washed) exhibited significantly enhanced responses towards dinotefuran over the concentration range of 10