EMAIL THIS PAGE TO A FRIEND
Chemico-biological interactions
Diquat-induced cellular pyridine nucleotide redox changes and alteration of metabolic enzyme activities in colonic carcinoma cells.
PMID 28108222
Abstract
Previously we have shown that the redox cycler menadione (MQ) induced cellular pyridine nucleotide redox imbalance that was linked to a decrease in aerobic glycolysis and perturbation of the mitochondrial respiratory activity due to the redox cycling of the compound; these processes were potentiated by low glucose. In this study, we investigated how colonic epithelial cells maintained pyridine nucleotide (NAD
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P13207
Phenazine, 98%
C12H8N2