EMAIL THIS PAGE TO A FRIEND
Free radical biology & medicine
Hydrogen sulfide reduces RAGE toxicity through inhibition of its dimer formation.
PMID 28108276
Abstract
RAGE is important in the development of neurodegenerative diseases. The present study was designed to investigate the effect of hydrogen sulfide (H