EMAIL THIS PAGE TO A FRIEND
Immunology letters
Thyroid hormone suppresses ischemia-reperfusion-induced liver NLRP3 inflammasome activation: Role of AMP-activated protein kinase.
PMID 28109981
Abstract
Thyroid hormone (T
Related Materials