EMAIL THIS PAGE TO A FRIEND
Parasitology international
Isolation and partial characterization of an immunogenic antigen of Giardia lamblia.
PMID 28110081
Abstract
Humoral and cellular immune responses play an important role during Giardia lamblia infection. Several Giardia proteins have been identified as immunogenic antigens based on their elicited humoral immune response. Poorly is known about Giardia antigens that stimulate a cellular immune response. The main purpose of this study was to isolate and partial characterize an immunogenic antigen (5G8) of G. lamblia. The 5G8 protein was isolated from G. lamblia trophozoite lysates by affinity chromatography using moAb 5G8-coupled CNBr-Sepharose. The isolated protein was analysed by electrospray tandem mass spectrometry (ESI-MS/MS), and by diverse bioinformatics tools (GiardiaDB, BLASTn, BLASTp and ExPASy). Additionally, several biochemical and immunological characteristics of the isolated protein were analysed. By ESI-MS/MS the amino acidic 5G8 sequence was deduced. The 5G8 antigen belongs to the VSP family proteins of G. lamblia. This protein is composed by one polypeptide chain (±71kDa). Using the algorithm SYFPHEITI, we identified candidate CD4
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
282332
Benzenesulfonyl fluoride, 99%
C6H5FO2S