Bulletin of environmental contamination and toxicology
Organochlorine Pesticides in Gonad, Brain, and Blood of Mice in Two Agricultural Areas of Sinaloa.
PMID 28110349
The adverse effect of pesticides on non-target wildlife and human health is a primary concern in the world, but in Mexico, we do not know which wildlife species are at the greatest risk. The aim of this study was to determine organochlorine pesticides in mice of two agricultural fields in Sinaloa, Culiacan and Guasave. Procedures of extraction, analysis, and quantification were followed according to the modified EPA 8081b method. In three mouse tissues (gonad, brain, and blood), γBHC and decachlorobiphenyl with a frequency higher than 50% and endosulfan sulfate with 43% were observed. The wildlife fauna living in agricultural areas are at great risk due to: (1) diversity of the chemicals used for pest control, like mice, and (2) variety of organochlorine pesticides in direct or indirect contact with non-target organisms, affecting the health of animals and humans (toxic effects and accumulation).