EMAIL THIS PAGE TO A FRIEND
Journal of materials science. Materials in medicine
Calcium-phosphate ceramics and polysaccharide-based hydrogel scaffolds combined with mesenchymal stem cell differently support bone repair in rats.
PMID 28110459
Abstract
Research in bone tissue engineering is focused on the development of alternatives to autologous bone grafts for bone reconstruction. Although multiple stem cell-based products and biomaterials are currently being investigated, comparative studies are rarely achieved to evaluate the most appropriate approach in this context. Here, we aimed to compare different clinically relevant bone tissue engineering methods and evaluated the kinetic repair and the bone healing efficiency supported by mesenchymal stem cells and two different biomaterials, a new hydrogel scaffold and a commercial hydroxyapatite/tricalcium phosphate ceramic, alone or in combination.Syngeneic mesenchymal stem cells (5 × 10