EMAIL THIS PAGE TO A FRIEND
Enzyme and microbial technology
Use of one- and two-mediator systems for developing a BOD biosensor based on the yeast Debaryomyces hansenii.
PMID 28110663
Abstract
We investigated the use of one- and two-mediator systems in amperometric BOD biosensors (BOD, biochemical oxygen demand) based on the yeast Debaryomyces hansenii. Screening of nine mediators potentially capable of electron transfer - ferrocene, 1,1'-dimethylferrocene, ferrocenecarboxaldehyde, ferroceneacetonitrile, neutral red, 2,6-dichlorophenolindophenol, thionine, methylene blue and potassium ferricyanide - showed only ferrocene and neutral red to be efficient electron carriers for the eukaryotes studied. Two-mediator systems based on combinations of the investigated compounds were used to increase the efficiency of electron transfer. The developed two-mediator biosensors exceeded their one-mediator analogs by their characteristics. The most preferable two-mediator system for developing a BOD biosensor was a ferrocene-methylene blue combination that ensured a satisfactory long-time stability (43 days), selectivity, sensitivity (the lower limit of the determined BOD5 concentrations, 2.5mg О2/dm(3)) and speed (assay time for one sample, not greater than 10min) of BOD determination. Analysis of water samples showed that the use of a ferrocene-methylene blue two-mediator system and the yeast D. hansenii enabled registration of data that highly correlated with the results of the standard method (R=0.9913).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
109576
1,1′-Dimethylferrocene, 95%
C12H14Fe
335053
Ferroceneacetonitrile
C12H11FeN