EMAIL THIS PAGE TO A FRIEND
Cellular immunology
miR-155 contributes to Df1-induced asthma by increasing the proliferative response of Th cells via CTLA-4 downregulation.
PMID 28110885
Abstract
Allergen-induced airway inflammation is characterized by Th2-mediated eosinophilic inflammation in the lungs. While the molecular mechanisms leading to this abnormal Th2 response remain unclear. Recent studies have demonstrated that MicroRNAs (miRNAs) modulate allergic airway inflammation. In this study, the role of miRNAs in allergic asthma pathogenesis was examined. Differentially expressed miRNAs were identified via miRNA microarray, with miR-155 being among the most highly expressed in asthma mice lungs. Examination of miR-155 overexpression resulted in enhanced inflammation and mucus hypersecretion in the lungs of allergen-challenged mice compared with control animals. Furthermore, CTLA-4, an important negative regulator of T-cell activation, was identified as a direct miR-155 target. Moreover, miR-155 overexpression in CD4
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
RAB0799 Mouse IgE ELISA Kit