EMAIL THIS PAGE TO A FRIEND
Gene
Mutations and interactions in human ERα and bZIP proteins: An in silico approach for cell signaling in breast oncology.
PMID 28111258
Abstract
I. Metastasis of breast cancer serves the most aggravating cause for transience in breast cancer patients. Accumulating evidences suggest that signal transduction in human breast cancers commences in estrogen-reliant pattern via signaling of the estrogen-receptor α-subunit (ERα) and XBP-1 (bZIP-domain) proteins. Furthermore, earlier investigations from SAGE and GST pull-down assay, also state that a point mutation in ERα leads to a risky factor by resulting into hyper-responsiveness towards estrogen and increased proliferation of breast cancer cells. So, a molecular-level exploration into the signaling mechanism is a prime requisite for future clinical and therapeutic progress. II. Present study explores primarily the residual participation of the two essential proteins from humans to boost the signaling mechanism in malignant breast tumors. So, 3D structures of the respective monomer proteins were demonstrated and mutated protein was homology modeled after the satisfaction of the stereo-chemical features. The functionality was observed to be conserved after mutation. Abrupt increment in protein-protein interactions was studied for the individual optimized and Molecular Dynamics simulated protein complexes. Revelation from supportive statistical significances for several energy calculations, solvent accessibility areas, electrostatic surface potentials and interaction studies led to confer that after mutation, the complex and the individual protein were the most stable and the best interactive one. For metastasis in breast cancer cells, polar charged residues hold a significant contribution. III. Therefore, this investigation provides a cogent framework for the interactive studies associated with breast cancer and an exposure towards the lethal impact on mutation.