EMAIL THIS PAGE TO A FRIEND
Molecular plant
Arabidopsis CBL-Interacting Protein Kinases Regulate Carbon/Nitrogen-Nutrient Response by Phosphorylating Ubiquitin Ligase ATL31.
PMID 28111287
Abstract
In response to the ratio of available carbon (C) and nitrogen (N) nutrients, plants regulate their metabolism, growth, and development, a process called the C/N-nutrient response. However, the molecular basis of C/N-nutrient signaling remains largely unclear. In this study, we identified three CALCINEURIN B-LIKE (CBL)-INTERACTING PROTEIN KINASES (CIPKs), CIPK7, CIPK12, and CIPK14, as key regulators of the C/N-nutrient response during the post-germination growth in Arabidopsis. Single-knockout mutants of CIPK7, CIPK12, and CIPK14 showed hypersensitivity to high C/low N conditions, which was enhanced in their triple-knockout mutant, indicating that they play a negative role and at least partly function redundantly in the C/N-nutrient response. Moreover, these CIPKs were found to regulate the function of ATL31, a ubiquitin ligase involved in the C/N-nutrient response via the phosphorylation-dependent ubiquitination and proteasomal degradation of 14-3-3 proteins. CIPK7, CIPK12, and CIPK14 physically interacted with ATL31, and CIPK14, acting with CBL8, directly phosphorylated ATL31 in a Ca