EMAIL THIS PAGE TO A FRIEND
Biochimie
Suppression of Neu1 sialidase delays the absorption of yolk sac in medaka (Oryzias latipes) accompanied with the accumulation of α2-3 sialo-glycoproteins.
PMID 28111290
Abstract
Sialidase catalyzes the removal of sialic acids from glycoconjugates. Recently, medaka sialidase Neu1 has been cloned and its enzymatic properties were investigated. Although enzymatic properties of this sialidase, such as optimal pH and substrate specificity, exhibits high similarity with human NEU1, Neu1 physiological functions in fish are still unclear. Here, to understand Neu1 significance in medaka embryogenesis, sialidase translation knockdown was carried out with one-cell stage fertilized egg using morpholino oligo injection. Neu1 exhibited desialylation of α2-3 sialic acid linkage in vitro and lysosomal localization in medaka caudal fin primary cells. Chloroquine treatment, inhibitor of lysosomal enzymes, caused an accumulation of α2-3 sialo-glycoproteins in the primary cells. During the embryogenesis neu1 mRNA level was elevated until 3.5 day post fertilization (dpf) while an initial decrease of α2-3 sialo-glycoprotein was observed around the same developmental stage. Neu1 knockdown by morpholino oligo induced some abnormal phenotypes such as delay of yolk sac absorption and small embryos. Sialidase-knockdown embryos also showed increase of heart rate in 5.5 and 6.5 dpf. Furthermore, about 37% decrease of hatching rate was observed in Neu1-MO treated embryos compared with control MO. Embryos showing severe phenotypes stopped embryogenesis at the late stage of development. Alteration of embryonic sialo-glycoproteins induced by morpholino injection was examined by lectin blotting to clarify the mechanism of abnormal development. As a result, degradation of several α2-3 sialo-glycoproteins was suppressed in Neu1-MO embryo, possibly induced by the interruption of lysosomal desialylation toward yolk glycoprotein. Our results suggest that medaka Neu1 could be crucial for embryonic development through the degradation of yolk sac nutrition.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I2149
Influenza Hemagglutinin (HA) Peptide, ≥97% (HPLC)
C53H67N9O17