EMAIL THIS PAGE TO A FRIEND
The international journal of biochemistry & cell biology
The selective cytotoxicity of DSF-Cu attributes to the biomechanical properties and cytoskeleton rearrangements in the normal and cancerous nasopharyngeal epithelial cells.
PMID 28111334
Abstract
Cancer initiation and progression follow complex changes of cellular architecture and biomechanical property. Cancer cells with more submissive (or "softer") than their healthy counterparts attributed to the reorganization of the complex cytoskeleton structure, may be considered as a potential anti-tumor therapeutic target. In this study, atomic force microscopy (AFM) was carried out to detect the topographical and biophysical changes of nasopharyngeal carcinoma CNE-2Z cells and normal nasopharyngeal epithelial cells NP69-SV40T by treating the Disulfiram chelated with Cu