Journal of cellular physiology
Intracellular transcytosis of albumin in glomerular endothelial cells after endocytosis through caveolae.
PMID 28112392
We previously described albumin endocytosis through caveolae in human renal glomerular endothelial cells (HRGECs). This suggested a new albumin transcytosis pathway, in addition to the fenestral pathway. As a next step, we investigated albumin transcytosis in HRGECs after caveolar endocytosis. HRGECs were incubated with Alexa Fluor 488-labeled bovine serum albumin from 0 to 360 min. Next, markers for endosomes, endoplasmic reticulum (ER), golgi apparatus (GA), lysosomes, and proteasomes and Fc receptors, microtubules, and actin were monitored by immunofluorescence. Labeled albumin co-localization with endosomes was gradually and significantly increased and it was significantly higher than with the other markers at any timepoint. Albumin, placed on inside of the Transwell membrane, diffused through HRGEC monolayers during a 360 min incubation period. This transportation of albumin through HRGECs was inhibited by methyl beta cyclodextrin (MBCD), a caveolae disrupting agent. MBCD also decreased albuminuria, causing decreased caveolin-1 (Cav-1) expression on glomerular capillaries, in puromycin aminonucleoside induced nephrotic mice. Albumin transcytosis depends on early endosomes, but not on other organelles, Fc receptors, or cytoskeletal components. Caveolae disruption prevented albumin transportation through HRGECs and decreased albuminuria in nephrotic mice. This newly described caveolae-dependent albumin pathway through glomerular endothelial cells is a potential pathogenetic mechanism for albuminuria, independent of the fenestrae.
Related Materials
Product #
Molecular Formula
Add to Cart
Puromycin aminonucleoside