EMAIL THIS PAGE TO A FRIEND
Biosensors & bioelectronics
Boronic Acid vs. Folic Acid: A Comparison of the bio-recognition performances by Impedimetric Cytosensors based on Ferrocene cored dendrimer.
PMID 28113133
Abstract
A comparative study is reported where folic acid (FA) and boronic acid (BA) based cytosensors and their analytical performances in cancer cell detection were analyzed by using electrochemical impedance spectroscopy (EIS) method. Cytosensors were fabricated using self-assembled monolayer principle by modifying Au electrode with cysteamine (Cys) and immobilization of ferrocene cored polyamidiamine dendrimers second generation (Fc-PAMAM (G2)), after which electrodes were modified with FA and BA. Au/Fc-PAMAM(G2)/FA and Au/Fc-PAMAM(G2)/BA based cytosensors showed extremely good analytical performances in cancer cell detection with linear range of 1×10
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CDS003827
3-Aminophenylboronic acid, AldrichCPR
C6H8BNO2
F408
Ferrocene, 98%
C10H10Fe