EMAIL THIS PAGE TO A FRIEND
Investigative ophthalmology & visual science
Role of Caveolin-1 for Blocking the Epithelial-Mesenchymal Transition in Proliferative Vitreoretinopathy.
PMID 28114583
Abstract
Proliferative vitreoretinopathy (PVR) is one of the most severe ocular diseases. Fibrotic changes in retinal cells are considered to be involved in the pathogenesis of PVR. Epithelial-mesenchymal transition (EMT) of RPE cells is one of the main concepts in the pathogenesis of fibrovascular membranes (FVMs) in PVR. In this study, we examined the expression of Caveolin-1 in human FVMs from patients with PVR. We also examined the role of Caveolin-1 in the pathogenesis of PVR. Western blotting, real-time PCR, and immunohistochemistry were performed with human FVMs and mouse eyes with PVR. Cell migration assays were performed to evaluate the involvement of Caveolin-1 in EMT using primary human and mouse RPE cells. Caveolin-1 was expressed in human FVMs and upregulated in the mouse eyes with PVR. The alpha-smooth muscle actin (αSMA) expression and migration ability were increased in RPE cells with knockout or knockdown of Caveolin-1, whereas zonula occludens-1 (ZO-1) immunohistochemistry showed reduced morphology and expression of ZO-1. In addition, migration assays showed that Caveolin-1 reduction increased RPE cell migration abilities. These results indicated that Caveolin-1 in RPE cells prevents PVR by blocking EMT.
Related Materials