EMAIL THIS PAGE TO A FRIEND
Carbohydrate polymers
Extraction of arabinoxylans from wheat bran using hydrothermal processes assisted by heterogeneous catalysts.
PMID 28115088
Abstract
The extraction/hydrolysis process of arabinoxylans from destarched wheat bran was studied in this work using different mesoporous silica supports and the corresponding RuCl
Related Materials