EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
The voltage-sensitive dye RH421 detects a Na
PMID 28115116
Abstract
RH421 is a voltage-sensitive fluorescent styrylpyridinium dye which has often been used to probe the kinetics of Na