EMAIL THIS PAGE TO A FRIEND
The Journal of neuroscience : the official journal of the Society for Neuroscience
Target Cell Type-Dependent Differences in Ca
PMID 28115484
Abstract
Target cell type-dependent differences in presynaptic release probability (