EMAIL THIS PAGE TO A FRIEND
The Journal of endocrinology
Esculentin-2CHa(1-30) and its analogues: stability and mechanisms of insulinotropic action.
PMID 28115493
Abstract
The insulin-releasing effects, cellular mechanisms of action and anti-hyperglycaemic activity of 10 analogues of esculentin-2CHa lacking the cyclic C-terminal domain (CKISKQC) were evaluated. Analogues of the truncated peptide, esculentin-2CHa(1-30), were designed for plasma enzyme resistance and increased biological activity. Effects of those analogues on insulin release, cell membrane integrity, membrane potential, intracellular Ca
Related Materials