EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
TMED10 Protein Interferes with Transforming Growth Factor (TGF)-β Signaling by Disrupting TGF-β Receptor Complex Formation.
PMID 28115518
Abstract
The intensity and duration of TGF-β signaling determine the cellular biological response. How this is negatively regulated is not well understood. Here, we identified a novel negative regulator of TGF-β signaling, transmembrane p24-trafficking protein 10 (TMED10). TMED10 disrupts the complex formation between TGF-β type I (also termed ALK5) and type II receptors (TβRII). Misexpression studies revealed that TMED10 attenuated TGF-β-mediated signaling. A 20-amino acid-long region from Thr