EMAIL THIS PAGE TO A FRIEND
Contact lens & anterior eye : the journal of the British Contact Lens Association
Activity of a melimine derived peptide Mel4 against Stenotrophomonas, Delftia, Elizabethkingia, Burkholderia and biocompatibility as a contact lens coating.
PMID 28118996
Abstract
To determine the antimicrobial activity of the melimine derived peptide Mel4 against Delftia, Stenotrophomonas, Elizabethkingia, Burkholderia and to investigate biocompatibility of Mel4 as an antimicrobial coating on contact lenses in animals and humans. In vitro antimicrobial activity of Mel4 was determined against the four Gram negative bacteria by investigating growth curves for 24h followed by viable counts to determine the minimum inhibitory concentration (MIC). Contact lenses were coated by covalently binding Mel4, characterized by amino acid analysis, and were investigated for changes in lens parameters. Safety of Mel-4 coated lenses were determined in a rabbit model of daily contralateral wear. A prospective, randomised, double-masked, contralateral, 1week daily wear human clinical trial was used to evaluate subjective responses and ocular physiology. Mel4 was active against all the bacteria tested (MIC Mel4 has achieved antimicrobial activity against variety of Gram negative bacteria that are often resistant to the action of cationic peptides and have been implicated in contact lens related adverse events. Mel4-coated contact lenses were safe to wear.