EMAIL THIS PAGE TO A FRIEND
World journal of microbiology & biotechnology
Cellulase with high β-glucosidase activity by Penicillium oxalicum under solid state fermentation and its use in hydrolysis of cassava residue.
PMID 28120306
Abstract
In this study, we investigated cellulase production by Penicillium oxalicum EU2106 under solid-state fermentation (SSF) and its hydrolysis efficiency toward NaOH-H