EMAIL THIS PAGE TO A FRIEND
Scientific reports
Mitochondrial and Chromosomal Damage Induced by Oxidative Stress in Zn
PMID 28120857
Abstract
Large-scale synthesis and release of nanomaterials in environment is a growing concern for human health and ecosystem. Therefore, we have investigated the cytotoxic and genotoxic potential of zinc oxide nanoparticles (ZnO-NPs), zinc oxide bulk (ZnO-Bulk), and zinc ions (Zn