EMAIL THIS PAGE TO A FRIEND
Biophysical journal
The Relaxation Properties of Myofibrils Are Compromised by Amino Acids that Stabilize α-Tropomyosin.
PMID 28122223
Abstract
We investigated the functional impact of α-tropomyosin (Tm) substituted with one (D137L) or two (D137L/G126R) stabilizing amino acid substitutions on the mechanical behavior of rabbit psoas skeletal myofibrils by replacing endogenous Tm and troponin (Tn) with recombinant Tm mutants and purified skeletal Tn. Force recordings from myofibrils (15°C) at saturating [Ca
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M0627
7-Methylguanosine, ≥90%
C11H15N5O5