EMAIL THIS PAGE TO A FRIEND
Journal of dentistry
Synthesis and characterization of a novel resin monomer with low viscosity.
PMID 28122255
Abstract
In this study, we designed and synthesized a novel macromolecule (tetramethyl bisphenol F acrylate, TMBPF-Ac) with low viscosity, excellent mechanical properties, and good biocompatibility. It could be used as a monomer for dental resin composites, which could reduce the risk of human exposure to bisphenol A derivatives in the oral environment. In addition, the monomer could be used without diluent, thereby avoiding the negative effect of a diluent METHODS: TMBPF-Ac was synthesized by a multistep condensation reaction. Its structure was confirmed by