Journal of Zhejiang University. Science. B
Isolation, chemical characterization, and immunomodulatory activity of naturally acetylated hemicelluloses from bamboo shavings.
PMID 28124842
Bamboo shavings, the outer or intermediate layer of bamboo stems, are the bulk of by-products produced in bamboo processing. In this study we investigated the isolation, chemical characterization, and immunostimulatory activity in vitro of the hemicelluloses from bamboo shavings. Shavings were first pretreated by steam explosion. The optimal pretreatment was found to be steam explosion at 2.2 MPa for 1 min. Following this pretreatment, the yield of hemicelluloses reached (2.05±0.22)% (based on the dry dewaxed raw materials), which was 5.7-fold higher than that of untreated samples. Bamboo-shavings hemicellulose (BSH) was then prepared by hot water extraction and ethanol precipitation from the steam-exploded shavings. Purification of BSH by anion-exchange chromatography of diethylaminoethanol (DEAE)-sepharose Fast Flow resulted in a neutral fraction (BSH-1, purity of 95.3%, yield of 1.06%) and an acidic fraction (BSH-2, purity of 92.5%, yield of 0.79%). The weight-average molecular weights (Mw) of BSH-1 and BSH-2 were 12 800 and 11 300 g/mol, respectively. Chemical and structural analyses by Fourier transform infrared spectroscopy (FT-IR), 1D ((1)H and (13)C) and 2D (heteronuclear single quantum correlation (HSQC)) nuclear magnetic resonance (NMR) spectra revealed that BSH-1 was O-acetylated-arabinoxylan and BSH-2 was O-acetylated-(4-O-methylglucurono)-arabinoxylan. BSH-1 had a higher content of acetyl groups than BSH-2. For the immunomodulatory activity in vitro, BSH and BSH-2 significantly stimulated mouse splenocyte proliferation while BSH-1 had no effect; BSH, BSH-1, and BSH-2 markedly enhanced the phagocytosis activity and nitric oxide production of the murine macrophage RAW264.7 in a dose-dependent manner. Our results suggest that the water-extractable hemicelluloses from steam-exploded bamboo shavings are naturally acetylated and have immunostimulatory activity.